ContactUsLinkComponent
Valved Respirator, FFP1
Valved Respirator, FFP1
Catalog # PRTWP101WHR
Supplier:  Portwest
Valved Respirator, FFP1
Catalog # PRTWP101WHR
Supplier:  Portwest
Specifications
  • Description:
    FFP1 valved respirator mask
  • Pk:
    10
Specifications
About this item
Ideal for applications involving dust, mists, and fibres, the P101 should be used to protect the wearer from solid, oil, and water-based aerosols.
  • Ultrasonic welded head straps
  • Foam padding around the nose bridge for additional comfort
  • High performance exhalation valve to reduce heat
  • Adjustable nose bar for improved fit and reduced fogging of glasses
  • Comfortable cup style respirator
  • Retail box aids presentation for retail sales
The exhalation valve expels heat and humidity from within the mask and reduces breathing resistance.
Certifications: EN 149:2001 +A1:2009 FFP1 (NR). CE-CAT III.