ContactUsLinkComponent
Pneumatic trough, pp, Nalgene®
  216-0428
   Low Manufacturing Impact
undefined
Pneumatic trough, pp, Nalgene®
  216-0428
   Low Manufacturing Impact
  • Description:
    Water
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
  • Pk:
    1
Replace metal-type troughs with the polypropylene pneumatic trough.
  • Lightweight and rustproof
  • Gas tubing passes through a loop to prevent liquid from flowing into gas line
  • Liquid inlet and outlet connections accept ¼" int.Ø × ³/₈" Ø (6,4×9,5 mm) tubing
This trough features tapered sides for nesting and a built-in gas inlet. There are no sharp corners or separate parts to lose. The built-in platform accommodates four 250 ml bottles.