Order Entry
Switzerland
ContactUsLinkComponent
(2,4-Dichlorophenoxy)acetic acid
Corrosive
Irritant
(2,4-Dichlorophenoxy)acetic acid
Catalog # 49083.
Supplier:  Merck
CAS Number:  
undefined
Corrosive
Irritant
(2,4-Dichlorophenoxy)acetic acid
Catalog # 49083.
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1
  • Seq:
    0001

Specifications

About this item

 :
 :221.04 g/mol
Merck Index:13,02825
Boiling Pt:346 °C (1013 hPa)
Melting Pt:137…139 °C
Density:1.563 g/cm³ (20 °C)
EINECS:202-361-1
 :GHS05
 :GHS07
Storage Temperature:Ambient
CAS Number:94-75-7
Formula:(C₆H₃Cl₂)OCH₂COOH
MDL Number:MFCD00004300