Supelco® Organic Reference Standard, 2-(4-Hydroxyphenyl)ethanol
Supplier: Merck
Certificates
About this item
- GHS signal: GHS07
Caution: P261 - P264 - P271 - P280 - P302 + P352 - P305 + P351 + P338
Specifications
- CAS No.:501-94-0
- Storage conditions:RT
- Hazard class description:H315 - H319 - H335
- Shipping conditions:STD
- MDL No.:MFCD00002902
- M-Clarity quality level:MQ100
- Synonym(s):4-Hydroxyphenethyl alcohol; Tyrosol; p-HPEA
- Melting point:89 - 92 °C (lit.); 90 - 93 °C
- Formula:HOC₆H₄CH₂CH₂OH
- Molecular weight:138,16
Specifications
Frequently Bought Together