Order Entry
Switzerland
ContactUsLinkComponent
 
 
 
 
1,3-Dimethyl-2-imidazolidinone ≥99.5% (by GC), CHROMASOLV™ for headspace gas chromatography, Riedel-de Haën™
 
 
 
 
1,3-Dimethyl-2-imidazolidinone ≥99.5% (by GC), CHROMASOLV™ for headspace gas chromatography, Riedel-de Haën™
 

 

  • Assay (GC):
    Min. 99.5 %
  • Refractive index (n 20/D):
    1.470 - 1.473
  • Water (Karl Fischer):
    Max. 0.04 %
  • Absorbance at 275 nm:
    Max. 0.522
  • Absorbance at 300 nm:
    Max. 0.221
  • Absorbance at 325 nm:
    Max. 0.096
  • Absorbance at 350 nm:
    Max. 0.045

 

 

Frequently Bought Together