Order Entry
Switzerland
ContactUsLinkComponent
Potassium dichromate concentrate for dilution to 1L, 1/60 M K2Cr2O7 (0.1N) volumetric solution, Fluka™
Corrosive
Health
Toxic
Potassium dichromate concentrate for dilution to 1L, 1/60 M K2Cr2O7 (0.1N) volumetric solution, Fluka™
Catalog # 38100-1EA
CAS Number:  
Corrosive
Health
Toxic
Potassium dichromate concentrate for dilution to 1L, 1/60 M K2Cr2O7 (0.1N) volumetric solution, Fluka™
Catalog # 38100-1EA
CAS Number:  

Specifications

  • Pack type:
    Ampoule
  • Pk:
    1
  • Seq:
    0001

Specifications

Specification Test Results

  • Factor:
    0.998 - 1.002

Specification Test Results

About this item

 :
 :294.18 g/mol
Boiling Pt:100 °C (1.013 hPa)
Density:1.030 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-906-6
UN:2922
ADR:8,II
 :GHS05
 :GHS08
 :GHS06
CAS Number:7778-50-9
Formula:K₂Cr₂O₇
MDL Number:MFCD00011367