Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Neutral red | Avantor

Neutral red

Glass bottle for solids, 10 g
Katalognr. ROTHT122.1
Marketsource
Anbieter:   Roth Carl
Neutral red
Katalognr. ROTHT122.1
Anbieter:  Roth Carl
Marketsource
CAS Nummer:  553-24-2
Neutral red

Glass bottle for solids, 10 g

CAS Nummer:  553-24-2
Preis
SFr. 110.30
Einloggen für Verfügbarkeit
1 * 10 g
Product Details & Documents
Synonyme:  N2,N2,7-Trimethylphenazine-2,8-diamine hydrochloride, C.I. 50040, 3-Amino-7-dimethyl amino-2-methylphenazine hydrochloride, N8,N8,3-Trimethyl-2,8-phenazinediamine monohydrochloride, Toluylene red, Basic Red 5
  • CAS Nummer:553-24-2
  • Formel:C₁₅H₁₇ClN₄
  • MDL-Nummer:MFCD00012651
  • Molekulargewicht:288.78 g/mol
  • Merck-Index:12,06571
  • Schmelzender Platingehalt:290 °C
  • Lagertemperatur:Ambient
  • EINECS:209-035-8
Technische Daten

Technische Daten

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g

Technische Daten

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.