Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Nitric acid
Nitric acid 1 mol/l (1 N) volumetric solution
Nitric acid 1 mol/l (1 N) volumetric solution
REGENO21001
:
REAGECON
:
Nitric acid 1 mol/l (1 N) volumetric solution
REGENO21001
:
REAGECON
:
Pack type:
Glass bottle
Pk:
1 L
Seq:
5037
:
:
63.01 g/mol
:
1.035 g/cm³ (25 °C)
:
Ambient
:
231-714-2
:
3264
:
8,III
:
GHS05
:
7697-37-2
:
HNO₃
:
MFCD00011349