Order Entry
Switzerland
ContactUsLinkComponent
Pepsin
Pepsin
Catalog # PROOFIP11
CAS Number:  
Pepsin
Catalog # PROOFIP11
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    2 g
  • CAS No.:
    9001-75-6
  • Storage conditions:
    Refrigerator
  • Enzyme name:
    Pepsin

Specifications

About this item