Order Entry
Switzerland
ContactUsLinkComponent
n-Pentane ≥95%, HiPerSolv CHROMANORM® for HPLC
Flammable
Health
Irritant
Environment
n-Pentane ≥95%, HiPerSolv CHROMANORM® for HPLC
Catalog # 83993.320
Supplier:  VWR Chemicals
CAS Number:  
undefined
Flammable
Health
Irritant
Environment
n-Pentane ≥95%, HiPerSolv CHROMANORM® for HPLC
Catalog # 83993.320
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L
  • Seq:
    5096

Specifications

Specification Test Results

  • Assay (on anhydrous substance):
    Min. 95.0 %
  • Acidity:
    Max. 0.0002 meq/g
  • Evaporation residue:
    Max. 5 ppm
  • Water:
    Max. 0.01 %
  • Transmittance (200 nm):
    Min. 56 %
  • Transmittance (210 nm):
    Min. 71 %
  • Transmittance (220 nm):
    Min. 89 %
  • Transmittance (230 nm):
    Min. 98 %
  • Transmittance (240 nm):
    Min. 99 %

Specification Test Results

About this item

 :
 :72.15 g/mol
Merck Index:13,07193
Boiling Pt:35…36 °C (1013 hPa)
Melting Pt:–130 °C
Density:0.63 g/cm³ (20 °C)
Flash Pt:–40 °C
Storage Temperature:Ambient
EINECS:203-692-4
UN:1265
ADR:3,II
 :Filtered through a 0.2 µm filter, packaged under nitrogen.
REACH:01-2119459286-30
 :GHS02
 :GHS08
 :GHS07
 :GHS09
CAS Number:109-66-0
Formula:H₃C(CH₂)₃CH₃
MDL Number:MFCD00009498