Order Entry
Switzerland
ContactUsLinkComponent
Nitric acid 6 mol/l (6 N), AVS TITRINORM volumetric solution
Corrosive
Toxic
Nitric acid 6 mol/l (6 N), AVS TITRINORM volumetric solution
Catalog # 310547.1000
Supplier:  VWR Chemicals
CAS Number:  
undefined
Corrosive
Toxic
Nitric acid 6 mol/l (6 N), AVS TITRINORM volumetric solution
Catalog # 310547.1000
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Titer (20°C; real value 0.2 % accuracy):
    6.00 N +/- 0,2%

Specification Test Results

About this item

 :
 :63.01 g/mol
Storage Temperature:Ambient
EINECS:231-714-2
UN:2031
ADR:8,II
 :GHS05
 :GHS06
CAS Number:7697-37-2
Formula:HNO₃
MDL Number:MFCD00011349