SFr. 0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Switzerland
ContactUsLinkComponent
4-Azaindole
4-Azaindole
4-Azaindole
4-Azaindole
Catalog # MOLEM21910645
Supplier:
Molekula
CAS Number:
Expand image
4-Azaindole
Catalog # MOLEM21910645
Supplier:
Molekula
CAS Number:
Specifications
Pack type:
Glass bottle for solids
Pk:
1 g
Seq:
3047
Specifications
About this item
Product Details
:
:
118.14 g/mol
CAS Number:
272-49-1
Formula:
C₇H₆N₂
MDL Number:
MFCD00971977
Product Details