Order Entry
Switzerland
ContactUsLinkComponent
Phthalaldehyde for synthesis, Sigma-Aldrich®
Phthalaldehyde for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Phthalaldehyde for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    53 g/l (20 °C)
  • Melting Point:
    54 - 58 °C
  • Molar Mass:
    134.13 g/mol
  • Bulk Density:
    530 kg/m3
  • Boiling Point:
    83 - 84 °C (1 hPa)
  • Flash Point:
    110 °C
  • Density:
    1.13 g/cm3 (20 °C)
  • pH:
    7 (53 g/l, H2O, 20 °C)
  • Ignition Point:
    480 °C
  • Hazard Class:
    8
  • Hazard Class Description:
    Corrosive material

Specification Test Results

Related Information

Frequently Bought Together