2-Bromethylamine hydrobromide for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: 2-Bromoethylammonium bromide, 2-Bromoethylamine hydrobromide
CAS Number:2576-47-8
:
Formula:BrCH₂CH₂NH₃Br
MDL Number:MFCD00012886
:204.89 g/mol
Melting Pt:170…173 °C
:M-Clarity™ quality level = MQ200
EINECS:219-924-2
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Solubility:500 g/l (20 °C)
- Melting Point:171 - 176 °C
- Molar Mass:204.89 g/mol
- pH:4.7 (25 g/l, H2O, 20 °C)
- Hazard Class:6.1
- Hazard Class Description:Toxic materials
Specification Test Results
Frequently Bought Together