

EDTA dipotassium salt dihydrate for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:25102-12-9
:
Formula:C₁₀H₁₄K₂N₂O₈·2H₂O
MDL Number:MFCD00150036
:404.46 g/mol
Merck Index:13,03546
Melting Pt:272 °C
Density:1 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
EINECS:217-895-0
:GHS08
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Solubility:(20 °C) soluble
- Melting Point:272 °C
- Molar Mass:404.47 g/mol
- Bulk Density:310 kg/m3
- Vapor Pressure:(20 °C)
- pH:4 - 5 (50 g/l, H2O, 20 °C)
Specification Test Results
Frequently Bought Together