


1,3-Dimethyl-2-imidazolidinone, Sigma-Aldrich®
: Merck
:80-73-9
:
:C₅H₁₀N₂O
:MFCD00003188
:114.15 g/mol
:13,03275
:225.5 °C (1013 hPa)
:8 °C
:1.04 g/cm³ (20 °C)
:201-304-8
:2810
:6.1,III
:M-Clarity™ quality level = MQ200
:93 °C
:GHS05
:GHS08
:GHS07
:Ambient
- Solubility:(20 °C) soluble
- Melting Point:8.2 °C
- Molar Mass:114.14 g/mol
- Boiling Point:104 °C (20 hPa)
- Vapor Pressure:- 1 hPa (20 °C)
- Flash Point:114 °C
- Refractive Index:1.472 (20 °C, 589 nm)
- Explosion Limit:1.3 - 8.4 %(V)
- Density:1.056 g/cm3 (20 °C)
- pH:(H2O) neutral
- Ignition Point:305 °C
- Synonyms:Dimethylethylene urea, DMI, DMEU
- Hill Formula:C5H10N2O
Frequently Bought Together