Ethyl benzoylacetate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:94-02-0
:
Formula:C₁₁H₁₂O₃
MDL Number:MFCD00009196
Merck Index:13,03803
Boiling Pt:265 °C (1013 hPa)
Melting Pt:< 0 °C
Density:1.114 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Flash Pt:140 °C
:192.21 g/mol
EINECS:202-295-3
Specification Test Results
- Solubility:(20 °C) insoluble
- Molar Mass:192.21 g/mol
- Boiling Point:265 - 270 °C (decomposition)
- Vapor Pressure:1 hPa (108 °C)
- Flash Point:140 °C
- Refractive Index:1.531 (20 °C, 589 nm)
- Density:1.114 g/cm3 (20 °C)
- Ignition Point:375 °C DIN 51794
- Synonyms:Benzoylacetic acid ethyl ester, 3-Oxo-3-phenylpropionic acid ethyl ester
- Hill Formula:C11H12O3
- Chemical Formula:C6H5COCH2COOC2H5
Specification Test Results
Frequently Bought Together