Order Entry
Switzerland
ContactUsLinkComponent
 
 
 
EDTA dipotassium salt dihydrate analytical reagent, Supelco®
 :  Merck
 
undefined
 
 
EDTA dipotassium salt dihydrate analytical reagent, Supelco®
 :  Merck
 

 

  • Solubility:
    (20 °C) soluble
  • Melting Point:
    272 °C
  • Molar Mass:
    404.47 g/mol
  • Bulk Density:
    310 kg/m3
  • Vapor Pressure:
    (20 °C)
  • pH:
    4 - 5 (50 g/l, H2O, 20 °C)

 

Frequently Bought Together