Order Entry
Switzerland
ContactUsLinkComponent
Formaldehyde ≥37% in aqueous solution stabilised USP, Ph. Eur., BP, SAFC®
Toxic
Health
Corrosive
Formaldehyde ≥37% in aqueous solution stabilised USP, Ph. Eur., BP, SAFC®
Supplier:  Merck
Certificates
Toxic
Health
Corrosive
Formaldehyde ≥37% in aqueous solution stabilised USP, Ph. Eur., BP, SAFC®
Supplier:  Merck
Certificates

Specification Test Results

  • Assay (iodometric):
    37.0 - 38.0 %
  • Identity:
    conforms
  • Appearance of solution:
    Passes test
  • Acidity (Ph Eur, BP):
    Passes test
  • Acidity (according to USP):
    Passes test

Specification Test Results

Related Information

Frequently Bought Together