


Phthalaldehyde ≥98%, yellow crystals
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:643-79-8
:
Formula:C₈H₆O₂
MDL Number:MFCD00003335
:134.13 g/mol
Boiling Pt:266 °C (1013 hPa)
Melting Pt:53.5…56.3 °C
Density:1.13 g/cm³ (20 °C)
Flash Pt:132 °C
Storage Temperature:Refrigerator
EINECS:211-402-2
UN:2923
ADR:6.1,II
:GHS05
:GHS06
:GHS07
Specification Test Results
- Identity Test:Passes
- Purity :≥98%
- FTIR:Conforms to Standard
- Melting point:53 - 58 °C
Specification Test Results
Frequently Bought Together