Neutral red
Glass bottle, 500 mL
Catalog # CTEH64065/5
Marketsource
Supplier: CLIN TECH
Jump to:
Product Details & Documents
Synonyms: N2,N2,7-Trimethylphenazine-2,8-diamine hydrochloride, C.I. 50040, 3-Amino-7-dimethyl amino-2-methylphenazine hydrochloride, N8,N8,3-Trimethyl-2,8-phenazinediamine monohydrochloride, Toluylene red, Basic Red 5
- CAS Number:553-24-2
- Formula:C₁₅H₁₇ClN₄
- MDL Number:MFCD00012651
- Molecular Weight:288.78 g/mol
- Merck Index:12,06571
- Melting Pt:290 °C
- Storage Temperature:Ambient
- EINECS:209-035-8
Specifications
Specifications
- Pack type:Glass bottle
- Pk:500 mL
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



