Order Entry
Switzerland
ContactUsLinkComponent
Pig Pepsin (from Gastric mucosa)
Pig Pepsin (from Gastric mucosa)
Catalog # CHMCAB1226
Supplier:  Chemicon
CAS Number:  
Pig Pepsin (from Gastric mucosa)
Catalog # CHMCAB1226
Supplier:  Chemicon
CAS Number:  

Specifications

  • Pk:
    25 mg
  • CAS No.:
    9001-75-6
  • Source:
    Gastric mucosa
  • Species:
    Pig
  • Storage conditions:
    Refrigerator
  • Enzyme name:
    Pepsin

Specifications

About this item