Order Entry
Switzerland
ContactUsLinkComponent
Methanol ≥99.5% (by GC) NF, Macron Fine Chemicals™
Flammable
Toxic
Health
Methanol ≥99.5% (by GC) NF, Macron Fine Chemicals™
Catalog # 8814-28
CAS Number:  
undefined
Flammable
Toxic
Health
Methanol ≥99.5% (by GC) NF, Macron Fine Chemicals™
Catalog # 8814-28
CAS Number:  

Specifications

  • Pack type:
    Steel drum
  • Pk:
    350 lb
  • Seq:
    7251

Specifications

Specification Test Results

  • Bulk Pharmaceutical Chemical:
    = 99.5 %
  • NF - Assay (CH₃OH) (by GC):
    ≥99.5 %
  • NF - Identification A:
    Passes Test
  • NF - Identification B:
    Passes Test
  • NF - Acidity:
    Passes Test
  • NF - Alkalinity (as NH₃):
    ≤3 ppm
  • NF - Nonvolatile Residue:
    ≤0.001 %
  • NF - Readily Carbonizable Substances:
    Passes Test
  • NF - Readily Oxidizable Substances:
    Passes Test
  • NF - Acetone, Aldehydes (as (CH₃)₂CO):
    ≤0.003 %
  • NF - Water (H₂O):
    ≤0.1 %

Specification Test Results

About this item

 :
 :32.04 g/mol
Merck Index:13,05984
Boiling Pt:64.6 °C (1013 hPa)
Melting Pt:–98 °C
Density:0.7918 g/cm³ (20 °C)
Flash Pt:11 °C (closed cup)
Storage Temperature:Ambient
EINECS:200-659-6
UN:1230
ADR:3,II
REACH:01-2119433307-44
 :GHS02
 :GHS06
 :GHS08
CAS Number:67-56-1
Formula:CH₃OH
MDL Number:MFCD00004595