Ethanol absolute, BAKER ANALYZED® HPLC for HPLC, analytical reagent, J.T.Baker®
: Avantor
:64-17-5
:
:H₃CCH₂OH
:MFCD00003568
:46.07 g/mol
:12,03806
:78.3 °C (1013 hPa)
:–117 °C
:0.7895 g/cm³ (20 °C)
:12 °C
:Ambient
:200-578-6
:1170
:3,II
:01-2119457610-43
:GHS02
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and After discussion
- Assay:min. 99.5 %
- Density (g/ml) at 20°C:0.789 - 0.790
- Free Acid (as CH3COOH):max. 0.001 %
- Residue after Evaporation:max. 0.001 %
- Water (H2O):max. 0.2 %
- at 210 nm:max. 0.7
- at 240 nm:max. 0.1
- at 260 nm:max. 0.01
Frequently Bought Together