Order Entry
Switzerland
ContactUsLinkComponent
Nitric acid 65%, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Oxidizing
Corrosive
Toxic
Nitric acid 65%, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Catalog # 6090.2500PE
Supplier:  Avantor
CAS Number:  
Oxidizing
Corrosive
Toxic
Nitric acid 65%, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Catalog # 6090.2500PE
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    2,5 L
  • Seq:
    5096

Specifications

Specification Test Results

  • Assay:
    min. 65 %
  • Dithizone determination:
    passes test
  • Chloride (Cl):
    max. 0.5 ppm
  • Mercury (Hg):
    max. 5 ppb
  • Residue after Ignition:
    max. 5 ppm
  • Sulfate (SO4):
    max. 1 ppm
  • Silver (Ag):
    max. 0.01
  • Aluminium (Al):
    max. 0.05
  • Arsenic (As):
    max. 0.05
  • Barium (Ba):
    max. 0.02
  • Beryllium (Be):
    max. 0.01
  • Calcium (Ca):
    max. 0.5
  • Cadmium (Cd):
    max. 0.02
  • Cobalt (Co):
    max. 0.01
  • Chromium (Cr):
    max. 0.1
  • Copper (Cu):
    max. 0.01
  • Iron (Fe):
    max. 0.1
  • Germanium (Ge):
    max. 0.05
  • Potassium (K):
    max. 0.1
  • Lithium (Li):
    max. 0.01
  • Magnesium (Mg):
    max. 0.1
  • Manganese (Mn):
    max. 0.01
  • Molybdenum (Mo):
    max. 0.02
  • Sodium (Na):
    max. 0.5
  • Nickel (Ni):
    max. 0.05
  • Lead (Pb):
    max. 0.05
  • Strontium (Sr):
    max. 0.01
  • Titanium (Ti):
    max. 0.1
  • Thallium (Tl):
    max. 0.05
  • Vanadium (V):
    max. 0.01
  • Zinc (Zn):
    max. 0.05
  • Zirconium (Zr):
    max. 0.1

Specification Test Results

About this item

 :
 :63.01 g/mol
Merck Index:13,06608
Boiling Pt:120.5 °C (1013 hPa)
Melting Pt:–42 °C
Density:1.4 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-714-2
UN:2031
ADR:8,II
 :GHS03
 :GHS05
 :GHS06
CAS Number:7697-37-2
Formula:HNO₃
MDL Number:MFCD00011349