Order Entry
Switzerland
ContactUsLinkComponent
Nitric acid concentrate for dilution to 1 mol in aqueous solution, DILUT-IT®, Laboratory Reagent, J.T.Baker®
Corrosive
Nitric acid concentrate for dilution to 1 mol in aqueous solution, DILUT-IT®, Laboratory Reagent, J.T.Baker®
Catalog # 4868.
Supplier:  Avantor
CAS Number:  
Corrosive
Nitric acid concentrate for dilution to 1 mol in aqueous solution, DILUT-IT®, Laboratory Reagent, J.T.Baker®
Catalog # 4868.
Supplier:  Avantor
CAS Number:  

Specifications

  • Pk:
    1
  • Seq:
    0001

Specifications

Specification Test Results

  • Titer (mol/l):
    0.998-1.002

Specification Test Results

About this item

 :
 :63.01 g/mol
Density:1.38 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-714-2
UN:2031
ADR:8,II
 :GHS05
CAS Number:7697-37-2
Formula:HNO₃
MDL Number:MFCD00011349