Order Entry
Switzerland
ContactUsLinkComponent
Sodium carbonate concentrate for dilution to 0.05 mol/l (0.1 N) in aqueous solution, DILUT-IT®, Laboratory Reagent, J.T.Baker®
Sodium carbonate concentrate for dilution to 0.05 mol/l (0.1 N) in aqueous solution, DILUT-IT®, Laboratory Reagent, J.T.Baker®
Catalog # 4683.
Supplier:  Avantor
CAS Number:  
undefined
Sodium carbonate concentrate for dilution to 0.05 mol/l (0.1 N) in aqueous solution, DILUT-IT®, Laboratory Reagent, J.T.Baker®
Catalog # 4683.
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Plastic ampoule
  • Pk:
    1 Ampoul
  • Seq:
    0501

Specifications

Specification Test Results

  • Titer (N):
    0.099-0.101

Specification Test Results

About this item

 :
 :105.99 g/mol
Storage Temperature:Ambient
EINECS:207-838-8
CAS Number:497-19-8
Formula:Na₂CO₃
MDL Number:MFCD00003494