Order Entry
Switzerland
ContactUsLinkComponent
 
Orthophosphoric acid 85 - 87% ≥85.0-87.0%, BAKER ANALYZED® ACS, J.T.Baker®
 :  Avantor
 
undefined
Orthophosphoric acid 85 - 87% ≥85.0-87.0%, BAKER ANALYZED® ACS, J.T.Baker®
 :  Avantor
 

 

  • Environmentally Preferable:

 

 

  • Meets ACS Reagent Chemical Requirements:
    85.0 - 87.0%
  • For Laboratory,Research,or Manufacturing Use:
    ≤0.002%
  • Assay (H₃PO₄) (by acidimetry):
    85.0 - 87.0 %
  • Calcium (Ca):
    ≤ 0.002 %
  • Color (APHA):
    ≤ 10
  • Insoluble Matter:
    ≤ 0.001 %
  • ACS - Magnesium (Mg):
    ≤ 0.002 %
  • Sulfate (SO₄):
    ≤ 12 ppm
  • Volatile Acids (as CH₃COOH):
    ≤ 0.001 %
  • Reducing Substances:
    Passes Test
  • Chloride (Cl):
    ≤ 3 ppm
  • Nitrate (NO₃):
    ≤ 5 ppm
  • Trace Impurities - Antimony (Sb):
    ≤ 20.000 ppm
  • Trace Impurities - Arsenic (As):
    ≤ 0.500 ppm
  • Trace Impurities - Iron (Fe):
    ≤ 10 ppm
  • Heavy Metals (as Pb):
    ≤ 8 ppm
  • Trace Impurities - Manganese (Mn):
    ≤ 0.500 ppm
  • Trace Impurities - Potassium (K):
    ≤ 40.000 ppm
  • Trace Impurities - Sodium (Na):
    ≤ 200.000 ppm

 

Frequently Bought Together