3-Methoxy-p-toluidine ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:16452-01-0
:
Formula:C₈H₁₁NO
MDL Number:MFCD00025371
Boiling Pt:250 °C (1013 hPa)
Melting Pt:56…60 °C
Storage Temperature:Ambient
EINECS:240-500-8
UN:2811
ADR:6.1,III
:137.18 g/mol
:GHS06
:GHS07
Related Information
Frequently Bought Together