3-Fluoro-p-anisidine ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:366-99-4
:
Formula:C₇H₈FNO
MDL Number:MFCD00075040
:141.15 g/mol
Boiling Pt:135 °C (18 mmHg)
Melting Pt:80…84 °C
Flash Pt:>110 °C (230 °F)
UN:2811
ADR:6.1,III
:GHS06
:GHS08
:GHS07
Storage Temperature:Ambient
Frequently Bought Together