

N,O-Bis(trimethylsilyl)trifluoroacetamide with 10% trimethylchlorosilane
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:25561-30-2
:
Formula:C₈H₁₈F₃NOSi₂
MDL Number:MFCD00008269
:257.4 g/mol
Flash Pt:23 °C (73 °F)
Storage Temperature:Refrigerator
EINECS:247-103-9
UN:2924
ADR:3,II
:GHS02
:GHS05
Related Information
Frequently Bought Together