
Dysprosium trichloride hydrate ≥99.99% (REO, rare earth oxide basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
:
Formula:Cl₃Dy·xH₂O
MDL Number:MFCD00010984
Melting Pt:163 °C
EINECS:233-039-9
:GHS07
Storage Temperature:Ambient
:268.86 g/mol (anhy)
Related Information
Frequently Bought Together