Order Entry
Switzerland
ContactUsLinkComponent
 
3-Fluoro-p-anisidine 99%
 
undefined
3-Fluoro-p-anisidine 99%

 

  • Appearance:
    Light beige to yellow-brown Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    81 to 84 °C
  • GC:
    ≥98.5 %

 

 

Frequently Bought Together