Order Entry
Switzerland
ContactUsLinkComponent
 
 
5-Hexyn-1-ol 97%
 
undefined
 
5-Hexyn-1-ol 97%

 

  • Appearance:
    Clear colorless to yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4470 to 1.4510 (20°C, 589 nm)
  • GC:
    ≥96.0 %

 

 

Frequently Bought Together