
di-Sodium succinate hexahydrate 97%
:6106-21-4
:
:NaOOCCH₂CH₂COONa·6H₂O
:MFCD00149117
:270.14 g/mol
:13,08754
:120 °C
:1,529 g/cm³ (20 °C)
:205-778-7
:GHS07
:Ambient
- Appearance:White Crystalline powder or crystals or granules
- Infrared spectrum:Conforms
- Loss on drying:37.0 % to 41.0 % (90°C)
- Solubility:(10 % in water) Clear
- Titration with HClO4:96.0 to 103.0 %
Frequently Bought Together