Order Entry
Switzerland
Orders LinkContactUsLinkComponent
cis-4-Hydroxy-D(+)-proline

cis-4-Hydroxy-D(+)-proline

Glass bottle for solids, 1 g
Catalog # MOLEM32560054
Marketsource
Supplier:   Molekula
cis-4-Hydroxy-D(+)-proline
Catalog # MOLEM32560054
Supplier:  Molekula
Marketsource
CAS Number:  2584-71-6
cis-4-Hydroxy-D(+)-proline

Glass bottle for solids, 1 g

CAS Number:  2584-71-6
Price
Inquire For Price
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  (R)-(+)-cis-4-Hydroxyproline, cis-4-Hydroxy-D-proline, H-D-cis-Hyp-OH, H-cis-D-Hyp-OH, D(+)-cis-4-Hydroxyproline
  • CAS Number:2584-71-6
  • Formula:C₅H₉NO₃
  • MDL Number:MFCD00005252
  • Molecular Weight:131.13 g/mol
  • EINECS:219-963-5
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.