cis-4-Hydroxy-D(+)-proline 99%
Supplier: Thermo Fisher Scientific
About this item
Synonyms: (R)-(+)-cis-4-Hydroxyproline, cis-4-Hydroxy-D-proline, H-D-cis-Hyp-OH, H-cis-D-Hyp-OH, D(+)-cis-4-Hydroxyproline
- CAS Number:2584-71-6
- Formula:C₅H₉NO₃
- MDL Number:MFCD00005252
- Molecular Weight:131.13 g/mol
- Melting Pt:243 °C (dec.)
- Storage Temperature:Ambient
- EINECS:219-963-5
2 Options Available Below
Product Details & Documents
Synonyms: (R)-(+)-cis-4-Hydroxyproline, cis-4-Hydroxy-D-proline, H-D-cis-Hyp-OH, H-cis-D-Hyp-OH, D(+)-cis-4-Hydroxyproline
- CAS Number:2584-71-6
- Formula:C₅H₉NO₃
- MDL Number:MFCD00005252
- Molecular Weight:131.13 g/mol
- Melting Pt:243 °C (dec.)
- Storage Temperature:Ambient
- EINECS:219-963-5
Documents
ACRO20491
SDS
Specifications for Product Family
Specification Test Results
- Appearance:White Powder
- Infrared spectrum:Conforms
- TLC:≥98.5 %
- Specific optical rotation:+56° to +60° (24°C, 589 nm) (c=2, water)
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



