Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

Ethanolamine 99%

Corrosive
Health
Irritant
Supplier:   Apollo Scientific
Ethanolamine 99%
Supplier:  Apollo Scientific
Certificates
Ethanolamine 99%

2 Options Available Below

Product Details & Documents
Synonyms:  2-Aminoethanol, Monoethanolamine
  • CAS Number:141-43-5
  • Formula:NH₂CH₂CH₂OH
  • MDL Number:MFCD00008183
  • Molecular Weight:61.08 g/mol
  • Boiling Pt:170 °C
  • Melting Pt:10…11 °C
  • Melting Pt:10 - 11 °C
  • Density:1.012
  • Flash Pt:91 °C
  • UN:2491
  • ADR:6.1,III
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - SFr. 26.00
    SFr. 26.00
    1 * 100 g
       SFr. 26.00

  • Bottle
    1 kg
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg - SFr. 122.00
    SFr. 122.00
    1 * 1 kg
       SFr. 122.00