Redox Sensors Memosens, CPS12E
Nome del fornitore: Endress+Hauser
New Product
Certificati
Informazioni su questo articolo
Digital all-round ORP sensor with low maintenance requirements and long service life.
- Robust, low-maintenance ORP sensor: PTFE junction prevents contamination by the medium; extended poison diffusion path avoids poisoning of the electrode reference
- Ideal for demanding environments: Gold or platinum components cover the entire oxidizing/reducing media; chemically stable shaft glass; pressure stability up to 17 bar absolute
- Memosens 2.0 for enhanced data storage: Enables advanced process/calibration data storage, enabling IIoT services and predictive maintenance
- Extremely reliable measurement through non-contact signal: Inductive transmission for increased process reliability
- Reduced downtime and costs: On-site laboratory calibration and fast sensor replacement minimize process interruptions and reduce operating costs
The Memosens CPS12E is a digital ORP sensor that ensures reliable measurements even in demanding environments or high-risk areas. Designed for minimal maintenance and long service life, it features Memosens 2.0 digital technology, which enables extended storage of calibration and process data, ideal for predictive maintenance. The use of Memosens' plug and play principle ensures straightforward operation and maximises process uptime.
Delivery: Digital ORP sensor Memosens CPS12E.
Frequently Bought Together