Order Entry
Switzerland
ContactUsLinkComponent
Organic reference standard, Carbon disulphide 5000 µg/ml in methanol, Supelco®
Organic reference standard, Carbon disulphide 5000 µg/ml in methanol, Supelco®
Katalognr. 40363.
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, Carbon disulphide 5000 µg/ml in methanol, Supelco®
Katalognr. 40363.
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1
  • CAS No.:
    75-15-0
  • Description:
    Carbon disulphide 5,000 µg/ml in methanol
  • Formula:
    CS₂
  • Description:
    Carbon disulphide

Specifications

About this item