Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Registrieren
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Heim
Cy3 aldehyde
Cy3 aldehyde for biomolecular labeling
Cy3 aldehyde for biomolecular labeling
Irritant
Cy3 aldehyde for biomolecular labeling
Catalog # AATB9010
Supplier:
AAT Bioquest
CAS Number:
Expand image
Irritant
Cy3 aldehyde for biomolecular labeling
Catalog # AATB9010
Supplier:
AAT Bioquest
CAS Number:
Specifications
Pack type:
Plastic bottle
Pk:
5 mg
Seq:
2210
Specifications
About this item
Product Details
:
:
919 g/mol
ADR:
5,II
:
GHS07
Product Details