
3-(4,4-difluoro-5,7-dimethyl-4-bora-3a,4a-diaza-s-indacene-3-yl)propionic acid NHS ester ≥90%
Catalog # AATB701
Supplier: AAT BIOQUEST
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:5 mg
- Seq:2210
Specifications
About this item
:
:389.166 g/mol
ADR:5,II
:GHS07
CAS Number:146616-66-2
Formula:C₁₈H₁₈BF₂N₃O₄
MDL Number:MFCD00467385