Order Entry
Switzerland
ContactUsLinkComponent
3'-DABCYL CPG 1000 Å for oligonucleotide labeling
Irritant
3'-DABCYL CPG 1000 Å for oligonucleotide labeling
Catalog # AATB6008
Supplier:  AAT Bioquest
CAS Number:  
undefined
Irritant
3'-DABCYL CPG 1000 Å for oligonucleotide labeling
Catalog # AATB6008
Supplier:  AAT Bioquest
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
ADR:5,II
 :GHS07