
Texas Red-X, succinimidyl ester single isomer fluorescent probe
Catalog # AATB474
Supplier: AAT Bioquest
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:5 mg
- Seq:2210
Specifications
About this item
:
:816.94 g/mol
ADR:5,II
:GHS07
CAS Number:216972-99-5
Formula:C₄₁H₄₄N₄O₁₀S₂