Order Entry
Switzerland
ContactUsLinkComponent
DSG (Disuccinimidyl glutarate) ≥90%
Reizend
DSG (Disuccinimidyl glutarate) ≥90%
Catalog # AATB4520
Anbieter:  AAT BIOQUEST
CAS-Nummer:  
undefined
Reizend
DSG (Disuccinimidyl glutarate) ≥90%
Catalog # AATB4520
Anbieter:  AAT BIOQUEST
CAS-Nummer:  

Technische Daten

  • Pack type:
    Plastic bottle
  • Pk:
    25 mg
  • Seq:
    2315

Technische Daten

Über diesen Artikel

 :
 :326.26 g/mol
ADR:5,II
 :GHS07
CAS-Nummer:79642-50-5
Formel:C₁₃H₁₄N₂O₈
MDL-Nummer:MFCD00153597