Sulpho-SMCC (3-Sulpho-N-succinimidyl-4-(N-maleimidomethyl)cyclohexane-1-carboxylate sodium salt) for peptide synthesis
Catalog # AATB4503
Supplier: AAT BIOQUEST
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:5 mg
- Seq:2210
Specifications
About this item
:
:436.37 g/mol
ADR:5,II
:GHS07
CAS Number:92921-24-9
Formula:C₁₆H₁₈N₂O₉S
MDL Number:MFCD00054978