Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Registrieren
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Heim
5-CR6G, SE
5-CR6G, SE single isomer ≥90%
5-CR6G, SE single isomer ≥90%
AATB345
:
AAT Bioquest
:
5-CR6G, SE single isomer ≥90%
AATB345
:
AAT Bioquest
:
Pk:
5 mg
Seq:
2210
:
:
555.59 g/mol
:
209112-21-0
:
C₃₁H₂₉N₃O₇