Order Entry
Switzerland
ContactUsLinkComponent
Biotin-PEG2-NHS biodetection
Irritant
Biotin-PEG2-NHS biodetection
Catalog # AATB3016
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
Irritant
Biotin-PEG2-NHS biodetection
Catalog # AATB3016
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    25 mg
  • Seq:
    2315

Specifications

About this item

 :
 :500.57 g/mol
ADR:5,II
 :GHS07
CAS Number:596820-83-6
 :MFCD11974976
Formula:C₂₁H₃₂N₄O₈S
MDL Number:MFCD00467154