
Trypan sodium salt 0.1 M, Trypan UltraBlue™ for dead cell labeling
Catalog # AATB2455
Supplier: AAT Bioquest
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:1 mL
- Seq:2070
Specifications
About this item
:
:1000 g/mol
ADR:5,II
:GHS07