SFr. 0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Switzerland
ContactUsLinkComponent
Tide Fluor™ 7WS alkyne
Tide Fluor™ 7WS alkyne, Tide Fluor™ fluorescent probe
Tide Fluor™ 7WS alkyne, Tide Fluor™ fluorescent probe
Irritant
Tide Fluor™ 7WS alkyne, Tide Fluor™ fluorescent probe
Catalog # AATB2305
Supplier:
AAT BIOQUEST
CAS Number:
Expand image
Irritant
Tide Fluor™ 7WS alkyne, Tide Fluor™ fluorescent probe
Catalog # AATB2305
Supplier:
AAT BIOQUEST
CAS Number:
Specifications
Pack type:
Plastic bottle
Pk:
1 mg
Seq:
2070
Specifications
About this item
Product Details
:
:
928.02 g/mol
ADR:
5,II
:
GHS07
Product Details